missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium oxide, 96%, extra pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 196910025

 View more versions of this product

Catalog No. AC196910025


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
Specifications

Specifications

2850.0°C
2570.0°C
Beige
2.5 kg
CaO
MFCD00010911
calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx
BRPQOXSCLDDYGP-UHFFFAOYSA-N
oxocalcium
14778
56.08
Extra Pure
Plastic bottle
Calcium oxide
1305-78-8
Powder
95% min. (Complexometry)
CaO
15, 1688
Solubility in water: reacts. Other solubilities: soluble in acids (reacts),soluble in glycerol,insoluble in alcohol
[O--].[Ca++]
56.08
CHEBI:31344
96%
3.5% max.

Chemical Identifiers

1305-78-8
56.08
BRPQOXSCLDDYGP-UHFFFAOYSA-N
14778
oxocalcium
CaO
MFCD00010911
calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx
CHEBI:31344
[O--].[Ca++]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 215-138-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only