Learn More
Calcium oxide, 96%, extra pure
CAS: 1305-78-8 | CaO | 56.08 g/mol
$243.99 - $464.93
Chemical Identifiers
| CAS | 1305-78-8 |
|---|---|
| Molecular Formula | CaO |
| Molecular Weight (g/mol) | 56.08 |
| MDL Number | MFCD00010911 |
| InChI Key | BRPQOXSCLDDYGP-UHFFFAOYSA-N |
| Synonym | calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx |
| PubChem CID | 14778 |
| ChEBI | CHEBI:31344 |
| IUPAC Name | oxocalcium |
| SMILES | [O--].[Ca++] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC196910010
|
Thermo Scientific Chemicals
196910010 |
1 kg | Glass bottle |
Each for $243.99
|
|
||||
|
AC196910025
|
Thermo Scientific Chemicals
196910025 |
2.5 kg | Plastic bottle |
Each for $464.93
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
AdsorbentChemical Identifiers
| 1305-78-8 | |
| 56.08 | |
| BRPQOXSCLDDYGP-UHFFFAOYSA-N | |
| 14778 | |
| oxocalcium |
| CaO | |
| MFCD00010911 | |
| calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx | |
| CHEBI:31344 | |
| [O--].[Ca++] |
Specifications
| 2850.0°C | |
| 1305-78-8 | |
| Powder | |
| 95% min. (Complexometry) | |
| CaO | |
| 15, 1688 | |
| Solubility in water: reacts. Other solubilities: soluble in acids (reacts), soluble in glycerol, insoluble in alcohol | |
| [O--].[Ca++] | |
| 56.08 | |
| CHEBI:31344 | |
| 96% | |
| 3.5% max. | |
| Calcium oxide |
| 2570.0°C | |
| Beige | |
| 1 kg | |
| CaO | |
| MFCD00010911 | |
| calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx | |
| BRPQOXSCLDDYGP-UHFFFAOYSA-N | |
| oxocalcium | |
| 14778 | |
| 56.08 | |
| Extra Pure | |
| Glass bottle |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 215-138-9
RUO – Research Use Only