missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Phosphate, Monobasic, Monohydrate, Crystal, BAKER ANALYZED™ Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
Supplier: Avantor J.T.Baker 142605
Specifications
| Calcium Phosphate | |
| 7758-23-8 | |
| 2.5 kg | |
| calcium biphosphate, monobasic calcium phosphate, calcium dihydrogen phosphate, acid calcium phosphate, primary calcium phosphate, monocalcium orthophosphate, c 38 phosphate, calcium diorthophosphate, monocalcium phosphate, monobasic, calcium dihydrogen orthophosphate | |
| OP(=O)(O)[O-].OP(=O)(O)[O-].[Ca+2] | |
| 234.05 | |
| CHEBI:35433 | |
| BAKER ANALYZED™ Reagent | |
| 1L = 2.22kg |
| Monobasic, Monohydrate | |
| Crystals | |
| CaH4O8P2 | |
| YYRMJZQKEFZXMX-UHFFFAOYSA-L | |
| calcium;dihydrogen phosphate | |
| 24454 | |
| 252.07 | |
| Poly Bottle |
Chemical Identifiers
| 7758-23-8 | |
| 234.05 | |
| calcium biphosphate, monobasic calcium phosphate, calcium dihydrogen phosphate, acid calcium phosphate, primary calcium phosphate, monocalcium orthophosphate, c 38 phosphate, calcium diorthophosphate, monocalcium phosphate, monobasic, calcium dihydrogen orthophosphate | |
| CHEBI:35433 | |
| OP(=O)(O)[O-].OP(=O)(O)[O-].[Ca+2] |
| CaH4O8P2 | |
| YYRMJZQKEFZXMX-UHFFFAOYSA-L | |
| 24454 | |
| calcium;dihydrogen phosphate |