missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium Phosphate, Monobasic, Monohydrate, Crystal, BAKER ANALYZED™ Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
$33238.78 - $37968.44
Chemical Identifiers
CAS | 7758-23-8 |
---|---|
Molecular Formula | CaH4O8P2 |
Molecular Weight (g/mol) | 234.05 |
InChI Key | YYRMJZQKEFZXMX-UHFFFAOYSA-L |
Synonym | calcium biphosphate, monobasic calcium phosphate, calcium dihydrogen phosphate, acid calcium phosphate, primary calcium phosphate, monocalcium orthophosphate, c 38 phosphate, calcium diorthophosphate, monocalcium phosphate, monobasic, calcium dihydrogen orthophosphate |
PubChem CID | 24454 |
ChEBI | CHEBI:35433 |
IUPAC Name | calcium;dihydrogen phosphate |
SMILES | OP(=O)(O)[O-].OP(=O)(O)[O-].[Ca+2] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
2002541
|
Avantor
142601 |
500 g | Poly Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
02002540
|
Avantor
142605 |
2.5 kg | Poly Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
7758-23-8 | |
234.05 | |
calcium biphosphate, monobasic calcium phosphate, calcium dihydrogen phosphate, acid calcium phosphate, primary calcium phosphate, monocalcium orthophosphate, c 38 phosphate, calcium diorthophosphate, monocalcium phosphate, monobasic, calcium dihydrogen orthophosphate | |
CHEBI:35433 | |
OP(=O)(O)[O-].OP(=O)(O)[O-].[Ca+2] |
CaH4O8P2 | |
YYRMJZQKEFZXMX-UHFFFAOYSA-L | |
24454 | |
calcium;dihydrogen phosphate |
Specifications
7758-23-8 | |
500 g | |
calcium biphosphate, monobasic calcium phosphate, calcium dihydrogen phosphate, acid calcium phosphate, primary calcium phosphate, monocalcium orthophosphate, c 38 phosphate, calcium diorthophosphate, monocalcium phosphate, monobasic, calcium dihydrogen orthophosphate | |
OP(=O)(O)[O-].OP(=O)(O)[O-].[Ca+2] | |
234.05 | |
CHEBI:35433 | |
BAKER ANALYZED™ Reagent | |
1L = 2.22kg |
Crystals | |
CaH4O8P2 | |
YYRMJZQKEFZXMX-UHFFFAOYSA-L | |
calcium;dihydrogen phosphate | |
24454 | |
252.07 | |
Poly Bottle | |
Calcium Phosphate, Monobasic, Monohydrate |