missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium phosphate tribasic, 34-40% Ca, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03673136

 View more versions of this product

Catalog No. AA3673136


Explore available promotions

Description

Description

Utilized in powdered spices as an anticaking agent and baby powder. It is the starting material for the production of phosphoric acid and fertilizers. Employed in tissue replacement for repairing bony defects and act as drug carrier for drug delivery to the bone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Calcium phosphate tribasic
White
Powder
34-40% Ca
MFCD00010904
Partially soluble in water.
[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
310.17
1004.67
3.14g/cm3 at 20°C
tribasic
12167-74-7
500 g
Ca3O8P2
durapatite, hydroxylapatite, hydroxyapatite, alveograf, periograf, ossopan, calcium hydroxyapatite, radiesse, monite, apatite, hydroxy
QORWJWZARLRLPR-UHFFFAOYSA-H
tricalcium diphosphate
14781
White powder

Chemical Identifiers

12167-74-7
310.17
QORWJWZARLRLPR-UHFFFAOYSA-H
14781
[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
Ca3O8P2
MFCD00010904
durapatite, hydroxylapatite, hydroxyapatite, alveograf, periograf, ossopan, calcium hydroxyapatite, radiesse, monite, apatite, hydroxy
tricalcium diphosphate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H315-H319-H335 : Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 235-330-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only