missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium phosphate tribasic, 34-40% Ca, Thermo Scientific Chemicals

$81.44 - $637.34

Chemical Identifiers

CAS 12167-74-7
Molecular Formula Ca3O8P2
Molecular Weight (g/mol) 310.17
MDL Number MFCD00010904
InChI Key QORWJWZARLRLPR-UHFFFAOYSA-H
Synonym durapatite, hydroxylapatite, hydroxyapatite, alveograf, periograf, ossopan, calcium hydroxyapatite, radiesse, monite, apatite, hydroxy
PubChem CID 14781
IUPAC Name tricalcium diphosphate
SMILES [Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3673136
View Documents
Thermo Scientific Chemicals
03673136
500 g
Each for $81.44
 
AA36731A3
View Documents
Thermo Scientific Chemicals
036731A3
2 kg
Each for $233.75
 
AA36731A9
View Documents
Thermo Scientific Chemicals
036731A9
10 kg
Each for $637.34
 
Description

Description

Utilized in powdered spices as an anticaking agent and baby powder. It is the starting material for the production of phosphoric acid and fertilizers. Employed in tissue replacement for repairing bony defects and act as drug carrier for drug delivery to the bone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12167-74-7
310.17
QORWJWZARLRLPR-UHFFFAOYSA-H
14781
[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
Ca3O8P2
MFCD00010904
durapatite, hydroxylapatite, hydroxyapatite, alveograf, periograf, ossopan, calcium hydroxyapatite, radiesse, monite, apatite, hydroxy
tricalcium diphosphate

Specifications

White
Powder
34-40% Ca
MFCD00010904
Partially soluble in water.
[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
310.17
1004.67
3.14g/cm3 at 20°C
12167-74-7
500 g
Ca3O8P2
durapatite, hydroxylapatite, hydroxyapatite, alveograf, periograf, ossopan, calcium hydroxyapatite, radiesse, monite, apatite, hydroxy
QORWJWZARLRLPR-UHFFFAOYSA-H
tricalcium diphosphate
14781
White powder
Calcium phosphate tribasic, tribasic
Videos
Safety and Handling

Safety and Handling

H315-H319-H335 : Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 235-330-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only