missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calmagite Indicator, 0.1% Aqueous, for Hardness, Certified, LabChem™

Supplier:  LabChem LC128501

 View more versions of this product

Catalog No. LC128501


Explore available promotions

Description

Description

Specifications

Specifications

Calmagite Indicator
3147-14-6,7732-18-5
C17H14N2O5S
Soluble in water
ASFVMSDYPYMUFL-UHFFFAOYSA-N
4-[(2-hydroxy-5-methylphenyl)hydrazinylidene]-3-oxonaphthalene-1-sulfonic acid
6364506
Certified
1g/mL
Nitrogen oxides; Carbon monoxide; Carbon dioxide
500 mL
0.1% Aqueous, for Hardness
99.9,0.1
C17H14N2O5S
Passes Test
CC1=CC(=C(C=C1)O)NN=C2C3=CC=CC=C3C(=CC2=O)S(=O)(=O)O
358.368
358.37
Poly Bottle
Passes Test
1g/mL
Liquid

Chemical Identifiers

3147-14-6
358.368
6364506
CC1=CC(=C(C=C1)O)NN=C2C3=CC=CC=C3C(=CC2=O)S(=O)(=O)O
C17H14N2O5S
ASFVMSDYPYMUFL-UHFFFAOYSA-N
4-[(2-hydroxy-5-methylphenyl)hydrazinylidene]-3-oxonaphthalene-1-sulfonic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions