missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calmagite Indicator, 0.1% Aqueous, for Hardness, Certified, LabChem™
$31.53 - $62.59
Chemical Identifiers
CAS | 3147-14-6 |
---|---|
Molecular Formula | C17H14N2O5S |
Molecular Weight (g/mol) | 358.368 |
InChI Key | ASFVMSDYPYMUFL-UHFFFAOYSA-N |
PubChem CID | 6364506 |
IUPAC Name | 4-[(2-hydroxy-5-methylphenyl)hydrazinylidene]-3-oxonaphthalene-1-sulfonic acid |
SMILES | CC1=CC(=C(C=C1)O)NN=C2C3=CC=CC=C3C(=CC2=O)S(=O)(=O)O |
Chemical Identifiers
3147-14-6 | |
358.368 | |
6364506 | |
CC1=CC(=C(C=C1)O)NN=C2C3=CC=CC=C3C(=CC2=O)S(=O)(=O)O |
C17H14N2O5S | |
ASFVMSDYPYMUFL-UHFFFAOYSA-N | |
4-[(2-hydroxy-5-methylphenyl)hydrazinylidene]-3-oxonaphthalene-1-sulfonic acid |
Specifications
3147-14-6,7732-18-5 | |
C17H14N2O5S | |
Soluble in water | |
ASFVMSDYPYMUFL-UHFFFAOYSA-N | |
4-[(2-hydroxy-5-methylphenyl)hydrazinylidene]-3-oxonaphthalene-1-sulfonic acid | |
6364506 | |
Certified | |
1g/mL | |
Nitrogen oxides; Carbon monoxide; Carbon dioxide | |
125 mL | |
Calmagite Indicator, 0.1% Aqueous, for Hardness |
99.9,0.1 | |
C17H14N2O5S | |
Passes Test | |
CC1=CC(=C(C=C1)O)NN=C2C3=CC=CC=C3C(=CC2=O)S(=O)(=O)O | |
358.368 | |
358.37 | |
Poly Bottle | |
Passes Test | |
1g/mL | |
Liquid |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature