missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cancer Diagnostics, Inc.™ Tissue Marking Dyes
Formulated to be used without a mordant
Supplier: Cancer Diagnostics, Inc.™ CDI07283
Description
- Dyes are pigment-based with an alcohol foundation for consistency and durability
- Proprietary blend of materials remains bright through processing and produces clean, crisp results
- Will not run or bleed on tissue
- Impervious to laboratory chemicals and reagents
Order Info
Individual colors available in two sizes; also available in systems with sets of six or seven colors, wood tray and 50 applicator sticks. Two-year shelf life.
Specifications
93°C | |
Green | |
Ea. | |
8.5 to 10 |
8 oz. | |
Tissue Marking Dye | |
Isopropyl Alcohol 400ppm, Ammonia 35ppm | |
Liquid |