missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cancer Diagnostics, Inc.™ Tissue Marking Dyes
Formulated to be used without a mordant
$141.03 - $1217.29
Specifications
Product Type | Tissue Marking Dye |
---|---|
Quantity | Ea. |
Formulation | Isopropyl Alcohol 400ppm, Ammonia 35ppm |
pH | 8.5 to 10 |
Form | Liquid |
Catalog Number | Mfr. No. | Color | Product Type | Capacity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Product Type | Capacity | Price | Quantity | |||
22500017
|
Cancer Diagnostics, Inc.™
CDI07276 |
Orange | Tissue Marking Dye | 2 oz. |
Each for $141.03
|
|
|||
22500014
|
Cancer Diagnostics, Inc.™
CDI07273 |
Green | Tissue Marking Dye | 2 oz. |
Each for $141.03
|
|
|||
22500031
|
Cancer Diagnostics, Inc.™
02000 |
Green, Black, Red, Yellow, Blue, Orange | Tissue Marking Dye | 2 oz. |
Pack of 6 Each for $1,000.65
|
|
|||
22500466
|
Cancer Diagnostics, Inc.™
CDI03000 |
Green, Black, Red, Yellow, Blue, Orange, Violet | Tissue Marking Dye | 2 oz. |
Pack of 7 Each for $1,217.29
|
|
|||
22500012
|
Cancer Diagnostics, Inc.™
07271 |
Black | Tissue Marking Dye | 2 oz. |
Each for $141.03
|
|
|||
22500013
|
Cancer Diagnostics, Inc.™
CDI07272 |
Blue | Tissue Marking Dye | 2 oz. |
Each for $141.03
|
|
|||
22500015
|
Cancer Diagnostics, Inc.™
CDI07274 |
Red | Tissue Marking Dye | 2 oz. |
Each for $141.03
|
|
|||
22500019
|
Cancer Diagnostics, Inc.™
07282 |
Blue | Tissue Marking Dye | 8 oz. |
Each for $361.94
|
|
|||
22500016
|
Cancer Diagnostics, Inc.™
07275 |
Yellow | Tissue Marking Dye | 2 oz. |
Each for $141.03
|
|
|||
22500018
|
Cancer Diagnostics, Inc.™
CDI07281 |
Black | Tissue Marking Dye | 8 oz. |
Each for $361.94
|
|
|||
22500022
|
Cancer Diagnostics, Inc.™
07285 |
Yellow | Tissue Marking Dye | 8 oz. |
Each for $361.94
|
|
|||
22500020
|
Cancer Diagnostics, Inc.™
CDI07283 |
Green | Tissue Marking Dye | 8 oz. |
Each for $361.94
|
|
|||
22500021
|
Cancer Diagnostics, Inc.™
07284 |
Red | Tissue Marking Dye | 8 oz. |
Each for $361.94
|
|
|||
22500023
|
Cancer Diagnostics, Inc.™
07286 |
Orange | Tissue Marking Dye | 8 oz. |
Each for $361.94
|
|
|||
Description
- Dyes are pigment-based with an alcohol foundation for consistency and durability
- Proprietary blend of materials remains bright through processing and produces clean, crisp results
- Will not run or bleed on tissue
- Impervious to laboratory chemicals and reagents
Order Info
Individual colors available in two sizes; also available in systems with sets of six or seven colors, wood tray and 50 applicator sticks. Two-year shelf life.
Specifications
Tissue Marking Dye | |
Isopropyl Alcohol 400ppm, Ammonia 35ppm | |
Liquid |
Ea. | |
8.5 to 10 |