missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BUCHI Capillaries for Melting Point Apparatus
For use with BUCHI Melting Point apparatus.
Supplier: BUCHI 001759
Description
- High-quality Borosilicate 3.3 glass
- Optimized thickness
- Reliable results
- Compatible with other automated instrument or manual techniques
Specifications
Melting Point Capillary | |
BUCHI Melting Point Apparatus | |
Glass |
180 x 120 x 80 mm | |
80 mm | |
1000 Tubes |