missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3M™ DBI-SALA™ ExoFit NEX™ Vest-Style Harness
Aluminum back D-ring, locking quick connect buckle leg straps, comfort padding.
Supplier: 3M™ 7012816115
Description
- Hybrid shoulder, back and leg comfort padding
- Stand-up Tech-Lite™ aluminum back D-ring
- Duo-Lok™ quick connect chest and leg straps
- Repel™ webbing
- Protective shoulder caps
- Loops for body belt
Specifications
Gray | |
4 lb. | |
Repel™ Polyester | |
Repel Polyester | |
ANSI A10.32, ANSI Z359.1, OSHA 1926.502, ANSI Z359.4, OSHA 1910.66 |
Vest-Style Harness | |
Large | |
6000 lb. | |
Back |