missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Capsaicin, USP, Spectrum™ Chemical
C1869, 404-86-4, C18H27NO3
Supplier: Spectrum Chemical Mfg Cor C18691KGBL
Description
Spectrum™ Chemical Capsaicin, USP is used as an analgesic in nasal spays, topical ointments and in dermal patches to relieve pain. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
404-86-4 | |
1% | |
57-66°C | |
C18H27NO3 | |
UN2811 | |
COC1=CC(CNC(=O)CCCC\C=C\C(C)C)=CC=C1O | |
305.42 | |
USP |
100% | |
1% | |
Amber Glass Bottle | |
1 kg | |
YKPUWZUDDOIDPM-SOFGYWHQSA-N | |
(6E)-N-[(4-hydroxy-3-methoxyphenyl)methyl]-8-methylnon-6-enamide | |
90 to 110% | |
Crystalline Powder or Lumps |
Chemical Identifiers
404-86-4 | |
305.42 | |
(6E)-N-[(4-hydroxy-3-methoxyphenyl)methyl]-8-methylnon-6-enamide |
C18H27NO3 | |
YKPUWZUDDOIDPM-SOFGYWHQSA-N | |
COC1=CC(CNC(=O)CCCC\C=C\C(C)C)=CC=C1O |