missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Capsaicin, USP, Spectrum™ Chemical
C1869, 404-86-4, C18H27NO3
$801.09 - $134399.37
Chemical Identifiers
CAS | 404-86-4 |
---|---|
Molecular Formula | C18H27NO3 |
Molecular Weight (g/mol) | 305.42 |
InChI Key | YKPUWZUDDOIDPM-SOFGYWHQSA-N |
IUPAC Name | (6E)-N-[(4-hydroxy-3-methoxyphenyl)methyl]-8-methylnon-6-enamide |
SMILES | COC1=CC(CNC(=O)CCCC\C=C\C(C)C)=CC=C1O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18601420
|
Spectrum Chemical Mfg Cor
C18691GM |
1 g | Amber Glass Bottle |
Each for $801.09
|
|
||||
18601421
|
Spectrum Chemical Mfg Cor
C18695GM |
5 g | Amber Glass Bottle |
Each for $2,847.23
|
|
||||
18601422
|
Spectrum Chemical Mfg Cor
C186925GM |
25 g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
18601423
|
Spectrum Chemical Mfg Cor
C1869100GMBL |
100 g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
18601424
|
Spectrum Chemical Mfg Cor
C18691KGBL |
1 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Capsaicin, USP is used as an analgesic in nasal spays, topical ointments and in dermal patches to relieve pain. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
404-86-4 | |
305.42 | |
(6E)-N-[(4-hydroxy-3-methoxyphenyl)methyl]-8-methylnon-6-enamide |
C18H27NO3 | |
YKPUWZUDDOIDPM-SOFGYWHQSA-N | |
COC1=CC(CNC(=O)CCCC\C=C\C(C)C)=CC=C1O |
Specifications
404-86-4 | |
1% | |
57-66°C | |
C18H27NO3 | |
UN2811 | |
COC1=CC(CNC(=O)CCCC\C=C\C(C)C)=CC=C1O | |
305.42 |
100% | |
1% | |
Amber Glass Bottle | |
1 g | |
YKPUWZUDDOIDPM-SOFGYWHQSA-N | |
(6E)-N-[(4-hydroxy-3-methoxyphenyl)methyl]-8-methylnon-6-enamide | |
USP |