missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Carbimazole, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 458610250

 View more versions of this product

Catalog No. AC458610250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Carbimazole
0.5% max. (24 h) (P2O5)
97.5% min. (HPLC)
25g
Solubility in water: 2g/L ( 20°C ). Other solubilities: soluble in acetone and ethanol
CCOC(=O)N1C=CN(C1=S)C
186.23
CHEBI:617099
98%
22232-54-8
Authentic
C7H10N2O2S
carbimazole, athyromazole, carbimazol, carbethoxymethimazole, neomercazole, carbinazole, neo-thyreostat, basolest, carbimazolum, thyrostat
CFOYWRHIYXMDOT-UHFFFAOYSA-N
ethyl 3-methyl-2-sulfanylideneimidazole-1-carboxylate
31072
186.23

Chemical Identifiers

22232-54-8
186.23
carbimazole, athyromazole, carbimazol, carbethoxymethimazole, neomercazole, carbinazole, neo-thyreostat, basolest, carbimazolum, thyrostat
CHEBI:617099
CCOC(=O)N1C=CN(C1=S)C
C7H10N2O2S
CFOYWRHIYXMDOT-UHFFFAOYSA-N
31072
ethyl 3-methyl-2-sulfanylideneimidazole-1-carboxylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May damage fertility or the unborn child.

GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Danger

EINECSNumber : 244-854-4

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only