missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Carbimazole, 98%, Thermo Scientific™

Chemical Identifiers

CAS 22232-54-8
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.23
InChI Key CFOYWRHIYXMDOT-UHFFFAOYSA-N
Synonym carbimazole, athyromazole, carbimazol, carbethoxymethimazole, neomercazole, carbinazole, neo-thyreostat, basolest, carbimazolum, thyrostat
PubChem CID 31072
ChEBI CHEBI:617099
IUPAC Name ethyl 3-methyl-2-sulfanylideneimidazole-1-carboxylate
SMILES CCOC(=O)N1C=CN(C1=S)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC458610050
View Documents
Thermo Scientific Chemicals
458610050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC458610250
View Documents
Thermo Scientific Chemicals
458610250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

22232-54-8
186.23
carbimazole, athyromazole, carbimazol, carbethoxymethimazole, neomercazole, carbinazole, neo-thyreostat, basolest, carbimazolum, thyrostat
CHEBI:617099
CCOC(=O)N1C=CN(C1=S)C
C7H10N2O2S
CFOYWRHIYXMDOT-UHFFFAOYSA-N
31072
ethyl 3-methyl-2-sulfanylideneimidazole-1-carboxylate

Specifications

22232-54-8
Authentic
C7H10N2O2S
carbimazole, athyromazole, carbimazol, carbethoxymethimazole, neomercazole, carbinazole, neo-thyreostat, basolest, carbimazolum, thyrostat
CFOYWRHIYXMDOT-UHFFFAOYSA-N
ethyl 3-methyl-2-sulfanylideneimidazole-1-carboxylate
31072
186.23
Carbimazole
0.5% max. (24 h) (P2O5)
97.5% min. (HPLC)
5g
Solubility in water: 2g/L ( 20°C ). Other solubilities: soluble in acetone and ethanol
CCOC(=O)N1C=CN(C1=S)C
186.23
CHEBI:617099
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May damage fertility or the unborn child.

GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Danger

EINECSNumber : 244-854-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only