Learn More
Carbon tetrabromide, 98%, Thermo Scientific Chemicals
$69.91 - $427.39
Chemical Identifiers
CAS | 558-13-4 |
---|---|
Molecular Formula | CBr4 |
Molecular Weight (g/mol) | 331.64 |
MDL Number | MFCD00000117 |
InChI Key | HJUGFYREWKUQJT-UHFFFAOYSA-N |
Synonym | carbon tetrabromide, methane, tetrabromo, carbon bromide, methane tetrabromide, bromid uhlicity, carbontetrabromide, methane, tetrabromide, tetrabrommethan, bromid uhlicity czech |
PubChem CID | 11205 |
ChEBI | CHEBI:47875 |
IUPAC Name | tetrabromomethane |
SMILES | C(Br)(Br)(Br)Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC325150050
|
Thermo Scientific Chemicals
325150050 |
5 g | Glass Bottle |
Each for $69.91
|
|
|||||
AC325151000
|
Thermo Scientific Chemicals
325151000 |
100 g | Plastic bottle |
Each for $128.94
|
|
|||||
AC325155000
|
Thermo Scientific Chemicals
325155000 |
500 g | Plastic bottle |
Each for $427.39
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
558-13-4 | |
331.64 | |
HJUGFYREWKUQJT-UHFFFAOYSA-N | |
11205 | |
tetrabromomethane |
CBr4 | |
MFCD00000117 | |
carbon tetrabromide, methane, tetrabromo, carbon bromide, methane tetrabromide, bromid uhlicity, carbontetrabromide, methane, tetrabromide, tetrabrommethan, bromid uhlicity czech | |
CHEBI:47875 | |
C(Br)(Br)(Br)Br |
Specifications
558-13-4 | |
White | |
97.5% min. (On organic basis) (GC) | |
CBr4 | |
MFCD00000117 | |
01, 68 | |
carbon tetrabromide, methane, tetrabromo, carbon bromide, methane tetrabromide, bromid uhlicity, carbontetrabromide, methane, tetrabromide, tetrabrommethan, bromid uhlicity czech | |
HJUGFYREWKUQJT-UHFFFAOYSA-N | |
tetrabromomethane | |
11205 | |
331.64 | |
Crystalline Solid |
88°C to 93°C | |
190°C | |
Glass Bottle | |
CBr4 | |
5 g | |
15,352; 16,366 | |
Solubility in water: insoluble | |
C(Br)(Br)(Br)Br | |
331.64 | |
CHEBI:47875 | |
98% | |
Carbon tetrabromide |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medi
GHS Signal Word: Warning
EINECSNumber : 209-189-6
RUO – Research Use Only