missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Carboxymethyl Cellulose Sodium Salt (n=approx. 500), TCI America™
$39.79 - $88.24
Chemical Identifiers
CAS | 9004-32-4 |
---|---|
Molecular Formula | (C12 H14 O9 R6)n |
Molecular Weight (g/mol) | 263.20 |
MDL Number | MFCD00081472 |
InChI Key | DPXJVFZANSGRMM-UHFFFAOYNA-N |
Synonym | carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn |
PubChem CID | 23706213 |
IUPAC Name | 2,3,4,5,6-pentahydroxyhexanal acetic acid sodium |
SMILES | [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O |
Chemical Identifiers
9004-32-4 | |
263.20 | |
DPXJVFZANSGRMM-UHFFFAOYNA-N | |
23706213 | |
[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O |
(C12 H14 O9 R6)n | |
MFCD00081472 | |
carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn | |
2,3,4,5,6-pentahydroxyhexanal acetic acid sodium |
Specifications
9004-32-4 | |
MFCD00081472 | |
carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn | |
[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O | |
263.20 | |
Crystalline Powder |
(C12 H14 O9 R6)n | |
25 g | |
DPXJVFZANSGRMM-UHFFFAOYNA-N | |
2,3,4,5,6-pentahydroxyhexanal acetic acid sodium | |
23706213 | |
Carboxymethyl Cellulose Sodium Salt (n=approx. 500) |
Safety and Handling
EINECSNumber : (8)-0203
RTECSNumber : FJ5950000
TSCA : Yes