Cardinal Health™ Regular (Flat) Sterility Maintenance Covers
Cardinal Health™ Sterility Maintenance Cover is used after sterilization to help maintain the sterility and cleanliness of the product.
Supplier: Cardinal Health™ T12430A
Description
Cardinal Health&trade Sterility Maintenance Cover is used after sterilization to help maintain the sterility and cleanliness of the product. The cover or bag is made of 3 mil low density polyethylene for strength with welded seals to help prevent seal opening in use. It includes a tear opening feature eliminating the need for scissors. The regular, flat cover can be heat sealed or taped closed after the contents are inserted.Specifications
Heat Seal or Tape | |
Polypropylene | |
30 in. | |
24 in. |
Ideal for covering wrapped items after sterilization to ensure maximum protection from bacteria, moisture and dust, can be heat sealed or taped | |
30 x 24 in. (76.2 x 61cm) | |
76.2 cm | |
61 cm |