Cardinal Health™ Regular (Flat) Sterility Maintenance Covers
Cardinal Health™ Sterility Maintenance Cover is used after sterilization to help maintain the sterility and cleanliness of the product.
$768.97 - $768.97
Specifications
Closure Type | Heat Seal or Tape |
---|---|
For Use With (Application) | Ideal for covering wrapped items after sterilization to ensure maximum protection from bacteria, moisture and dust, can be heat sealed or taped |
Material | Polypropylene |
Catalog Number | Mfr. No. | Length (Metric) | Width (Metric) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Width (Metric) | Price | Quantity | ||||
19-160-154
|
Cardinal Health™
T12430A |
76.2 cm | 61 cm |
Case of 250 Each for $768.97
|
|
||||
Description
Cardinal Health™ Sterility Maintenance Cover is used after sterilization to help maintain the sterility and cleanliness of the product. The cover or bag is made of 3 mil low density polyethylene for strength with welded seals to help prevent seal opening in use. It includes a tear opening feature eliminating the need for scissors. The regular, flat cover can be heat sealed or taped closed after the contents are inserted.Specifications
Heat Seal or Tape | |
Ideal for covering wrapped items after sterilization to ensure maximum protection from bacteria, moisture and dust, can be heat sealed or taped | |
Polypropylene |