Promega CellTiter-Glo™ 3D Cell Viability Assay
The CellTiter-Glo™ 3D Cell Viability Assay is a homogeneous method to determine the number of viable cells in 3D cell culture based on quantitation of the ATP present, which is a marker for the presence of metabolically active cells.
$252.97 - $1371.63
Description
- A Cell Viability Assay Validated for 3D Microtissue Cultures
- Accurate 3D cytotoxicity determination
- Easy assay implementation
- Simple, 30-minute protocol
Specifications
-30°C to -10°C |