Promega CellTiter-Glo™ 3D Cell Viability Assay DFS Item
The CellTiter-Glo™ 3D Cell Viability Assay is a homogeneous method to determine the number of viable cells in 3D cell culture based on quantitation of the ATP present, which is a marker for the presence of metabolically active cells.
Supplier:  Promega G9681
 View more versions of this product
Catalog No. PRG9681
Explore available promotions
Description
Description
  • A Cell Viability Assay Validated for 3D Microtissue Cultures
  • Accurate 3D cytotoxicity determination
  • Easy assay implementation
  • Simple, 30-minute protocol
Specifications
Specifications
10 mL
-30°C to -10°C