CEP Haz Mat-Aggressive Sorbent Spill Kits
For fast hazardous spill response
Supplier:  CEP HAZSK20
 View more versions of this product
Catalog No. 19-056-215
Explore available promotions
Description
Description
Spill kits include sorbents listed in table plus goggles, gloves, and disposable bag within a plastic drum.
Specifications
Specifications
20 gal.
Hazmat Aggressive Sorbent Spill Kit
20 gal. lab pack with lid, 50 sorbent pads, 4 sorbent socks, 2 sorbent pillows, 1 pair of goggles, 1 pair of gloves, 1 disposable bag
75.7 L
Fast hazardous spill response
Product Suggestions
Product Suggestions