missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
CEP Haz Mat-Aggressive Sorbent Spill Kits
For fast hazardous spill response
$194.61 - $359.82
Specifications
Type | Hazmat Aggressive Sorbent Spill Kit |
---|---|
For Use With (Application) | Fast hazardous spill response |
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Includes | Price | Quantity | ||||
19-056-215
|
CEP
HAZSK20 |
20 gal. | 75.7 L | 20 gal. lab pack with lid, 50 sorbent pads, 4 sorbent socks, 2 sorbent pillows, 1 pair of goggles, 1 pair of gloves, 1 disposable bag |
Each for $359.82
|
|
||||
19-056-217
|
CEP
HAZSK5 |
5 gal. | 18.92 L | 5 gal. plastic pail with lid, 10 sorbent pads, 2 sorbent socks, 1 pair of goggles, 1 pair of gloves, 1 disposable bag |
Each for $194.61
|
|
||||
Description
Spill kits include sorbents listed in table plus goggles, gloves, and disposable bag within a plastic drum.
Specifications
Hazmat Aggressive Sorbent Spill Kit | |
Fast hazardous spill response |