missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ceric Ammonium Sulfate, 0.100 N (N/10) in 1 N Sulfuric Acid, Ricca Chemical

Supplier:  Ricca Chemical Company 189016

 View more versions of this product

Catalog No. 189016


Explore available promotions

Description

Description

Specifications

Specifications

Ceric Ammonium Sulfate
10378-47-9,7732-18-5,7664-93-9
5.77,91.0,5.03
Ceric ammonium sulfate
N.N.N.N.O.O.[Ce+4].OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O
632.53
Natural Poly Bottle
500 mL
0.100N
Orange
5.55,88.0,4.84
CeH20N4O18S4
VCNAMBGKEDPVGQ-UHFFFAOYSA-J
λ⁴-cerium(4+) tetraamine dihydrate tetrahydrogen sulfate
16211593
1N Sulfuric Acid
Liquid

Chemical Identifiers

10378-47-9
632.53
Ceric ammonium sulfate
λ⁴-cerium(4+) tetraamine dihydrate tetrahydrogen sulfate
CeH20N4O18S4
VCNAMBGKEDPVGQ-UHFFFAOYSA-J
16211593
N.N.N.N.O.O.[Ce+4].OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O
Videos
SDS Documents

Documents

Promotions

Promotions