missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ceric Ammonium Sulfate, 0.100 N (N/10) in 1 N Sulfuric Acid, Ricca Chemical

$143.41 - $497.50

Chemical Identifiers

CAS 10378-47-9
Molecular Formula CeH20N4O18S4
Molecular Weight (g/mol) 632.53
InChI Key VCNAMBGKEDPVGQ-UHFFFAOYSA-J
Synonym Ceric ammonium sulfate
PubChem CID 16211593
IUPAC Name λ⁴-cerium(4+) tetraamine dihydrate tetrahydrogen sulfate
SMILES N.N.N.N.O.O.[Ce+4].OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
189016
SDS View Documents
Ricca Chemical Company
189016
500 mL Natural Poly Bottle
Each for $143.41
 
189032
View Documents
Ricca Chemical Company
189032
1 L Natural Poly Bottle
Each for $225.00
 
18901
View Documents
Ricca Chemical Company
18901
4 L Natural Poly Bottle
Each for $497.50
 
Description

Description

Specifications

Chemical Identifiers

10378-47-9
632.53
Ceric ammonium sulfate
λ⁴-cerium(4+) tetraamine dihydrate tetrahydrogen sulfate
CeH20N4O18S4
VCNAMBGKEDPVGQ-UHFFFAOYSA-J
16211593
N.N.N.N.O.O.[Ce+4].OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O

Specifications

Orange
5.55,88.0,4.84
CeH20N4O18S4
VCNAMBGKEDPVGQ-UHFFFAOYSA-J
λ⁴-cerium(4+) tetraamine dihydrate tetrahydrogen sulfate
16211593
1N Sulfuric Acid
Liquid
Ceric Ammonium Sulfate
10378-47-9,7732-18-5,7664-93-9
5.77,91.0,5.03
Ceric ammonium sulfate
N.N.N.N.O.O.[Ce+4].OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O
632.53
Natural Poly Bottle
500 mL
0.100N
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'