missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ceric Sulfate, 0.100 N (N/10) in 2 N Sulfuric Acid, Ricca Chemical

Supplier:  Ricca Chemical Company 19001

 View more versions of this product

Catalog No. 1900-1


Explore available promotions

Description

Description

Specifications

Specifications

Ceric Sulfate
13590-82-4,7664-93-9,7732-18-5
MFCD00148852
VZDYWEUILIUIDF-UHFFFAOYSA-J
λ⁴-cerium(4+) disulfate
159684
2N Sulfuric Acid
Liquid
Orange
CeO8S2
ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate
[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
332.23
Natural Poly Bottle
4 L
0.100N

Chemical Identifiers

13590-82-4
332.23
VZDYWEUILIUIDF-UHFFFAOYSA-J
159684
[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
CeO8S2
MFCD00148852
ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate
λ⁴-cerium(4+) disulfate
Videos
SDS
Documents

Documents

Promotions

Promotions