missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ceric Sulfate, 0.100 N (N/10) in 2 N Sulfuric Acid, Ricca Chemical

$177.00 - $616.13

Chemical Identifiers

CAS 13590-82-4
Molecular Formula CeO8S2
Molecular Weight (g/mol) 332.23
MDL Number MFCD00148852
InChI Key VZDYWEUILIUIDF-UHFFFAOYSA-J
Synonym ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate
PubChem CID 159684
IUPAC Name λ⁴-cerium(4+) disulfate
SMILES [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
1900-16
View Documents
Ricca Chemical Company
190016
500 mL Natural Poly Bottle
Each for $177.00
 
1900-32
SDS View Documents
Ricca Chemical Company
190032
1 L Natural Poly Bottle
Each for $251.78
 
1900-1
View Documents
Ricca Chemical Company
19001
4 L Natural Poly Bottle
Each for $616.13
 
Description

Description

Specifications

Chemical Identifiers

13590-82-4
332.23
VZDYWEUILIUIDF-UHFFFAOYSA-J
159684
[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
CeO8S2
MFCD00148852
ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate
λ⁴-cerium(4+) disulfate

Specifications

Orange
CeO8S2
ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate
[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
332.23
Natural Poly Bottle
500 mL
0.100N
13590-82-4,7664-93-9,7732-18-5
MFCD00148852
VZDYWEUILIUIDF-UHFFFAOYSA-J
λ⁴-cerium(4+) disulfate
159684
2N Sulfuric Acid
Liquid
Ceric Sulfate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'