missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ceric Sulfate, 0.100 N (N/10) in 2 N Sulfuric Acid, Ricca Chemical
Supplier: Ricca Chemical Company 190016
Specifications
Ceric Sulfate | |
13590-82-4,7664-93-9,7732-18-5 | |
MFCD00148852 | |
VZDYWEUILIUIDF-UHFFFAOYSA-J | |
λâ´-cerium(4+) disulfate | |
159684 | |
2N Sulfuric Acid | |
Liquid |
Orange | |
CeO8S2 | |
ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate | |
[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
332.23 | |
Natural Poly Bottle | |
500 mL | |
0.100N |
Chemical Identifiers
13590-82-4 | |
332.23 | |
VZDYWEUILIUIDF-UHFFFAOYSA-J | |
159684 | |
[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
CeO8S2 | |
MFCD00148852 | |
ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate | |
λâ´-cerium(4+) disulfate |