missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium, 99.95+%, (trace metal basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 219190010

 View more versions of this product

Catalog No. AC219190010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Specifications

Golden
99.95+%
7440-46-2
1.8730g/mL
Cs
1 g
caesium, cesium-133, cesium, elemental, unii-1ksv9v4y4i, metals basis, 1ksv9v4y4i, cesio, zaesium, casium, zasium
[Cs]
132.91
CHEBI:30514
≥99.95%
Cesium
28.5°C
705.0°C
99.95+%
MFCD00134037
15, 2007
TVFDJXOCXUVLDH-UHFFFAOYSA-N
cesium
5354618
132.91
Trace Metal Basis

Chemical Identifiers

7440-46-2
132.91
TVFDJXOCXUVLDH-UHFFFAOYSA-N
5354618
[Cs]
Cs
MFCD00134037
caesium, cesium-133, cesium, elemental, unii-1ksv9v4y4i, metals basis, 1ksv9v4y4i, cesio, zaesium, casium, zasium
CHEBI:30514
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.

GHS P Statement:
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 231-155-4

RTECSNumber : FK9225000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only