missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium carbonate, 99.5%, for analysis, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 192040250

 View more versions of this product

Catalog No. AC192040250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cesium carbonate
99.5%
White
25 g
CCs2O3
MFCD00010957
15,2009
Solubility in water: 261 g/100mL water (20°C). Other solubilities: soluble in alcohol and ether
[Cs+].[Cs+].[O-]C([O-])=O
325.82
325.82
Analytical
Plastic bottle
4.0720g/mL
>400°C
534-17-8
Crystalline Powder
99.4% min. (By difference)
Cs2CO3
07,57; 08,81; 09,99; 12,108; 13,70; 14,77; 15,73
cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate
FJDQFPXHSGXQBY-UHFFFAOYSA-L
dicesium;carbonate
10796
99.5%
0.5% max. (500°C)
4.072

Chemical Identifiers

534-17-8
325.82
FJDQFPXHSGXQBY-UHFFFAOYSA-L
10796
CCs2O3
MFCD00010957
cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate
[Cs+].[Cs+].[O-]C([O-])=O
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of damaging fertility or the unborn child.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure if swallowed.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

GHS Signal Word: Danger

EINECSNumber : 208-591-9

RTECSNumber : FK9400000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only