Learn More
Cesium carbonate, 99.5%, for analysis, Thermo Scientific Chemicals
$37.12 - $893.97
Chemical Identifiers
CAS | 534-17-8 |
---|---|
Molecular Formula | CCs2O3 |
Molecular Weight (g/mol) | 325.82 |
MDL Number | MFCD00010957 |
InChI Key | FJDQFPXHSGXQBY-UHFFFAOYSA-L |
Synonym | cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate |
PubChem CID | 10796 |
SMILES | [Cs+].[Cs+].[O-]C([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC192040050
|
Thermo Scientific Chemicals
192040050 |
5 g | Plastic bottle |
N/A
|
|
||||
AC192040250
|
Thermo Scientific Chemicals
192040250 |
25 g | Plastic bottle |
N/A
|
|
||||
AC192041000
|
Thermo Scientific Chemicals
192041000 |
100 g | Plastic bottle |
N/A
|
|
||||
AC192045000
|
Thermo Scientific Chemicals
192045000 |
500 g | Plastic bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
534-17-8 | |
325.82 | |
FJDQFPXHSGXQBY-UHFFFAOYSA-L | |
10796 |
CCs2O3 | |
MFCD00010957 | |
cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate | |
[Cs+].[Cs+].[O-]C([O-])=O |
Specifications
>400°C | |
White | |
5 g | |
CCs2O3 | |
MFCD00010957 | |
15,2009 | |
Solubility in water: 261 g/100mL water (20°C). Other solubilities: soluble in alcohol and ether | |
[Cs+].[Cs+].[O-]C([O-])=O | |
325.82 | |
325.82 | |
Analytical | |
Plastic bottle | |
4.0720g/mL |
534-17-8 | |
Crystalline Powder | |
99.4% min. (By difference) | |
Cs2CO3 | |
07,57; 08,81; 09,99; 12,108; 13,70; 14,77; 15,73 | |
cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate | |
FJDQFPXHSGXQBY-UHFFFAOYSA-L | |
dicesium;carbonate | |
10796 | |
99.5% | |
0.5% max. (500°C) | |
4.072 | |
Cesium carbonate, 99.5% |
Safety and Handling
GHS H Statement
Suspected of damaging fertility or the unborn child.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure if swallowed.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
GHS Signal Word: Danger
EINECSNumber : 208-591-9
RTECSNumber : FK9400000
TSCA : TSCA
RUO – Research Use Only