missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium fluoride, 99.9%, extra pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 315910050

 View more versions of this product

Catalog No. AC315910050

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Specifications

1251.0°C
682.0°C
White
5 g
CsF
MFCD00010960
15, 2011
Solubility in water: 369g/100mL (18°C). Other solubilities: soluble in methanol, insoluble in dioxane and pyridine
[F-].[Cs+]
151.90
151.91
Extra Pure
Cesium fluoride
13400-13-0
Powder
99.9%
CsF
01,121; 06,100; 07,57; 08,81; 09,100; 10,81; 11,115; 12,108; 13,68; 14,79; 15,75; 16,69; 17,68
cesium fluoride, caesium fluoride, cesium monofluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride
XJHCXCQVJFPJIK-UHFFFAOYSA-M
caesium(1+) fluoride
25953
99.9%
Plastic bottle

Chemical Identifiers

13400-13-0
151.90
XJHCXCQVJFPJIK-UHFFFAOYSA-M
25953
CsF
MFCD00010960
cesium fluoride, caesium fluoride, cesium monofluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride
[F-].[Cs+]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic in contact with skin.
Causes serious eye damage.
Causes skin irritation.
Toxic if inhaled.
Toxic if swallowed.
Suspected of damaging fertility or the unborn child if inhaled.
Contact with a

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISO

GHS Signal Word: Danger

EINECSNumber : 236-487-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only