Learn More
Cesium fluoride, 99.9%, extra pure, Thermo Scientific Chemicals
$76.80 - $161.72
Chemical Identifiers
CAS | 13400-13-0 |
---|---|
Molecular Formula | CsF |
Molecular Weight (g/mol) | 151.90 |
MDL Number | MFCD00010960 |
InChI Key | XJHCXCQVJFPJIK-UHFFFAOYSA-M |
Synonym | cesium fluoride, caesium fluoride, cesium monofluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride |
PubChem CID | 25953 |
SMILES | [F-].[Cs+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC315910050
|
Thermo Scientific Chemicals
315910050 |
5 g | Plastic bottle |
Each for $76.80
|
|
||||
AC315910250
|
Thermo Scientific Chemicals
315910250 |
25 g | Plastic bottle |
Each for $161.72
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
13400-13-0 | |
151.90 | |
XJHCXCQVJFPJIK-UHFFFAOYSA-M | |
25953 |
CsF | |
MFCD00010960 | |
cesium fluoride, caesium fluoride, cesium monofluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride | |
[F-].[Cs+] |
Specifications
1251.0°C | |
13400-13-0 | |
Powder | |
99.9% | |
CsF | |
01,121; 06,100; 07,57; 08,81; 09,100; 10,81; 11,115; 12,108; 13,68; 14,79; 15,75; 16,69; 17,68 | |
cesium fluoride, caesium fluoride, cesium monofluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride | |
XJHCXCQVJFPJIK-UHFFFAOYSA-M | |
caesium(1+) fluoride | |
25953 | |
99.9% | |
Plastic bottle |
682.0°C | |
White | |
5 g | |
CsF | |
MFCD00010960 | |
15, 2011 | |
Solubility in water: 369g/100mL (18°C). Other solubilities: soluble in methanol, insoluble in dioxane and pyridine | |
[F-].[Cs+] | |
151.90 | |
151.91 | |
Extra Pure | |
Cesium fluoride |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Causes serious eye damage.
Causes skin irritation.
Toxic if inhaled.
Toxic if swallowed.
Suspected of damaging fertility or the unborn child if inhaled.
Contact with a
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISO
GHS Signal Word: Danger
EINECSNumber : 236-487-3
RUO – Research Use Only