missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium iodide, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 192820010

 View more versions of this product

Catalog No. AC192820010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Specifications

1280.0°C
626.0°C
White
1 g
CsI
MFCD00010962
cesium iodide, caesium iodide, cesium monoiodide, dicesium diiodide, tricesium triiodide, cesiumiodide, cesium iodide cs2i2, cesium iodide cs3i3, unii-u1p3gvc56l
XQPRBTXUXXVTKB-UHFFFAOYSA-M
caesium(1+) iodide
24601
99.999%
Glass bottle
Cesium iodide
7789-17-5
Crystalline Powder
99.999%
CsI
15,2013
Solubility in water: 740g/L (20°C). Other solubilities: soluble in ethanol, slightly soluble in methanol, practically insoluble in acetone
[I-].[Cs+]
259.81
259.8
Trace Metal Basis

Chemical Identifiers

7789-17-5
259.81
XQPRBTXUXXVTKB-UHFFFAOYSA-M
24601
CsI
MFCD00010962
cesium iodide, caesium iodide, cesium monoiodide, dicesium diiodide, tricesium triiodide, cesiumiodide, cesium iodide cs2i2, cesium iodide cs3i3, unii-u1p3gvc56l
[I-].[Cs+]
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of damaging fertility or the unborn child if inhaled.
Very toxic to aquatic life.

GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the

GHS Signal Word: Warning

EINECSNumber : 232-145-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only