Learn More
Cesium iodide, 99.999%, (trace metal basis), Thermo Scientific Chemicals
$49.41 - $157.12
Chemical Identifiers
CAS | 7789-17-5 |
---|---|
Molecular Formula | CsI |
Molecular Weight (g/mol) | 259.81 |
MDL Number | MFCD00010962 |
InChI Key | XQPRBTXUXXVTKB-UHFFFAOYSA-M |
Synonym | cesium iodide, caesium iodide, cesium monoiodide, dicesium diiodide, tricesium triiodide, cesiumiodide, cesium iodide cs2i2, cesium iodide cs3i3, unii-u1p3gvc56l |
PubChem CID | 24601 |
SMILES | [I-].[Cs+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC192820010
|
Thermo Scientific Chemicals
192820010 |
1 g | Glass bottle |
Each for $49.41
|
|
||||
AC192820100
|
Thermo Scientific Chemicals
192820100 |
10 g | Glass bottle |
Each for $157.12
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
7789-17-5 | |
259.81 | |
XQPRBTXUXXVTKB-UHFFFAOYSA-M | |
24601 |
CsI | |
MFCD00010962 | |
cesium iodide, caesium iodide, cesium monoiodide, dicesium diiodide, tricesium triiodide, cesiumiodide, cesium iodide cs2i2, cesium iodide cs3i3, unii-u1p3gvc56l | |
[I-].[Cs+] |
Specifications
1280.0°C | |
7789-17-5 | |
Crystalline Powder | |
99.999% | |
CsI | |
15,2013 | |
Solubility in water: 740g/L (20°C). Other solubilities: soluble in ethanol, slightly soluble in methanol, practically insoluble in acetone | |
[I-].[Cs+] | |
259.81 | |
259.8 | |
Trace Metal Basis | |
Cesium iodide |
626.0°C | |
White | |
1 g | |
CsI | |
MFCD00010962 | |
cesium iodide, caesium iodide, cesium monoiodide, dicesium diiodide, tricesium triiodide, cesiumiodide, cesium iodide cs2i2, cesium iodide cs3i3, unii-u1p3gvc56l | |
XQPRBTXUXXVTKB-UHFFFAOYSA-M | |
caesium(1+) iodide | |
24601 | |
99.999% | |
Glass bottle |
Safety and Handling
GHS H Statement
Suspected of damaging fertility or the unborn child if inhaled.
Very toxic to aquatic life.
GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the
GHS Signal Word: Warning
EINECSNumber : 232-145-2
RUO – Research Use Only