CHEMetrics COD Vials Kits
Contains premeasured reagents for accurate, economical determination of COD
Supplier: CHEMetrics K7370S
Description
- Contains everything needed (except distilled water) to perform tests
- Includes vials and instruction book
- Comes in cardboard box
- 2-hour digestion time
- Dichromate reactor digestion
A fresh reagent ampule blank must be prepared for each series of tests; therefore the number of samples that can be tested with each kit will vary.
Certifications
Not USEPA Approved
Specifications
Chemical Oxygen Demand (COD) | |
Not USEPA approved | |
Digestor block as well as a photometer or spectrophotometer | |
24 | |
Dichromate Reactor Digestion | |
COD Vials Kit |
Not USEPA approved | |
Premeasured reagents for accurate, economical determination of COD, determination of the oxidizable organic matter content of wastewater | |
25 vials and instruction book | |
0 to 15,000 ppm (HR+) | |
2 hr. digestion |