missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
CHEMetrics COD Vials Kits
Contains premeasured reagents for accurate, economical determination of COD
$132.79 - $715.65
Specifications
Analytes | Chemical Oxygen Demand (COD) |
---|---|
For Use With (Application) | Premeasured reagents for accurate, economical determination of COD, determination of the oxidizable organic matter content of wastewater |
For Use With (Equipment) | Digestor block as well as a photometer or spectrophotometer |
Test Time | 2 hr. digestion |
Type | COD Vials Kit |
Includes: Instructions
Catalog Number | Mfr. No. | Certifications/Compliance | Range | No. of Tests | Includes | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Certifications/Compliance | Range | No. of Tests | Includes | Price | Quantity | ||
13-299-730
|
CHEMetrics
K7350S |
USEPA accepted | 0 to 150 ppm | 24 | 25 vials and instruction book |
Each for $149.76
|
|
||
36-100-7032
|
CHEMetrics
K7355 |
USEPA accepted | 0 to 150 ppm | 149 | 150 vials and instruction book |
Each for $663.12
|
|
||
13-299-732
|
CHEMetrics
K7360S |
USEPA accepted | 0 to 1500 ppm | 25 | 25 vials and instruction book |
Each for $132.79
|
|
||
13-299-230
|
CHEMetrics
K7365 |
USEPA accepted | 0 to 1500 ppm | 149 | 150 vials and instruction book |
Pack of 150 Each for $715.65
|
|
||
13-299-437
|
CHEMetrics
K7370S |
Not USEPA approved | 0 to 15,000 ppm (HR+) | 24 | 25 vials and instruction book |
Each for $137.82
|
|
||
Description
- Contains everything needed (except distilled water) to perform tests
- Includes vials and instruction book
- Comes in cardboard box
- 2-hour digestion time
- Dichromate reactor digestion
A fresh reagent ampule blank must be prepared for each series of tests; therefore the number of samples that can be tested with each kit will vary.
Specifications
Chemical Oxygen Demand (COD) | |
Digestor block as well as a photometer or spectrophotometer | |
COD Vials Kit |
Premeasured reagents for accurate, economical determination of COD, determination of the oxidizable organic matter content of wastewater | |
2 hr. digestion |