CHEMetrics COD Vials Kits
Contains premeasured reagents for accurate, economical determination of COD
Supplier:  CHEMetrics K7360S
 View more versions of this product
Catalog No. 13-299-732
Explore available promotions
Description
Description
  • Contains everything needed (except distilled water) to perform tests
  • Includes vials and instruction book
  • Comes in cardboard box
  • 2-hour digestion time
  • Dichromate reactor digestion
A fresh reagent ampule blank must be prepared for each series of tests; therefore the number of samples that can be tested with each kit will vary.
Specifications
Specifications
Chemical Oxygen Demand (COD)
Premeasured reagents for accurate, economical determination of COD, determination of the oxidizable organic matter content of wastewater
25 vials and instruction book
0 to 1500 ppm
2 hr. digestion
USEPA accepted
Digestor block as well as a photometer or spectrophotometer
25
Dichromate Reactor Digestion
COD Vials Kit