missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloramphenicol Crystalline MP Biomedicals

Supplier:  MP Biomedicals Inc 0219032191

 View more versions of this product

Catalog No. ICN19032191


Explore available promotions

Description

Description

Specifications

Specifications

Chloramphenicol
1.586 g/mL
1 kg
WIIZWVCIJKGZOK-RKDXNWHRSA-N
2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-yl]acetamide
5959
323.15
56-75-7
C11H12Cl2N2O5
chloramphenicol, chlornitromycin, chloromycetin, levomycetin, chlorocid, halomycetin, levomicetina, alficetyn, fenicol, globenicol
C1=CC(=CC=C1C(C(CO)NC(=O)C(Cl)Cl)O)[N+](=O)[O-]
323.126
CHEBI:17698
Crystalline

Chemical Identifiers

56-75-7
323.126
chloramphenicol, chlornitromycin, chloromycetin, levomycetin, chlorocid, halomycetin, levomicetina, alficetyn, fenicol, globenicol
CHEBI:17698
C1=CC(=CC=C1C(C(CO)NC(=O)C(Cl)Cl)O)[N+](=O)[O-]
C11H12Cl2N2O5
WIIZWVCIJKGZOK-RKDXNWHRSA-N
5959
2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-yl]acetamide
Videos
SDS
Documents

Documents

Promotions

Promotions