missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroacetone, 96%, stabilized, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 173472500

 View more versions of this product

Catalog No. AC173472500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Chloroacetone
78-95-5
Brown-Yellow to Yellow
119.0°C
Authentic
Glass bottle
1.4310 to 1.4330
MFCD00000936
01, 653
15, 2114
Solubility in water: 124g/L (20°C). Other solubilities: soluble in alcohol, ether and chloroform
CC(=O)CCl
92.52
6571
CHEBI:47220
96%
Stabilized
-44.5°C
1.1600g/mL
32°C
95% min. (GC)
C3H5ClO
ClCH2COCH3
250 mL
1.16
chloroacetone, chloropropanone, acetonyl chloride, monochloroacetone, tonite, 2-propanone, 1-chloro, chloro-2-propanone, 1-chloroacetone, chloromethyl methyl ketone, 1-chloro-2-propanone
BULLHNJGPPOUOX-UHFFFAOYSA-N
1-chloropropan-2-one
0.1% epoxidized soybean oil
1.11 mPa.s (25°C)
92.52
Liquid

Chemical Identifiers

78-95-5
92.52
BULLHNJGPPOUOX-UHFFFAOYSA-N
6571
1-chloropropan-2-one
C3H5ClO
MFCD00000936
chloroacetone, chloropropanone, acetonyl chloride, monochloroacetone, tonite, 2-propanone, 1-chloro, chloro-2-propanone, 1-chloroacetone, chloromethyl methyl ketone, 1-chloro-2-propanone
CHEBI:47220
CC(=O)CCl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic in contact with skin.
Causes serious eye irritation.
Toxic if swallowed.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Fatal if inhaled.
Causes ski

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre

GHS Signal Word: Danger

EINECSNumber : 201-161-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only