missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroform (Ethanol as Preservative/Certified ACS), Fisher Chemical™

Meet or exceeds the latest ACS specifications. Actual lot of analysis on label

Supplier:  Fisher Chemical BPC2981

 View more versions of this product

Catalog No. C298-1


Explore available promotions

Description

Description

Specifications

Specifications

Chloroform
Approx. 0.75% ethanol as preservative
Colorless
1.480g/cm³
1 L
Glass Bottle
0.001% max.
CHCl3
213mbar at 20°C
0.02% max.
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane
6212
119.38
Certified ACS Reagent
-63°C
67-66-3,64-17-5
290°C
61°C
Pass Test
Liquid
≥99.8 %
MFCD00000826
0.56 mPa/s at 20°C
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
ClC(Cl)Cl
119.37
CHEBI:35255
≥99.8%
0.75% Ethanol

Chemical Identifiers

67-66-3
119.37
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
trichloromethane
CHCl3
MFCD00000826
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
CHEBI:35255
ClC(Cl)Cl
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. Irritating to eyes and skin. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. May be harmful if inhaled. May cause irritation of respiratory tract. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.

NFPA

Health:2
Flammability:1
Instability:0

DOTInformation : DOT Class 6.1, : Poison

SDS Documents

Documents

Promotions

Promotions