Learn More
Chloroform (Ethanol as Preservative/Certified ACS), Fisher Chemical™
Meet or exceeds the latest ACS specifications. Actual lot of analysis on label
$257.91 - $33034.09
Chemical Identifiers
| CAS | 67-66-3 |
|---|---|
| Molecular Formula | CHCl3 |
| Molecular Weight (g/mol) | 119.37 |
| MDL Number | MFCD00000826 |
| InChI Key | HEDRZPFGACZZDS-UHFFFAOYSA-N |
| Synonym | trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio |
| PubChem CID | 6212 |
| ChEBI | CHEBI:35255 |
| IUPAC Name | trichloromethane |
| SMILES | ClC(Cl)Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
C298-500
|
Thermo Fisher Scientific
BPC298500 |
500 mL | Glass Bottle |
Each for $257.91
Case of 6 Each for $1,547.46
|
|
||||
|
C298-1
|
Thermo Fisher Scientific
BPC2981 |
1 L | Glass Bottle |
Case of 6 Each for $1,510.09
|
|
||||
C298-4
|
Fisher Chemical
BPC2984 |
4 L | Glass Bottle |
Each for $812.27
Case of 4 Each for $3,249.08
|
|
||||
|
C298-20
|
Fisher Chemical
BP29820 |
20 L | Steel Pail |
Each for $3,357.55
|
|
||||
|
C298-200
|
Fisher Chemical
BPC298200 |
200 L | Steel Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 67-66-3 | |
| 119.37 | |
| HEDRZPFGACZZDS-UHFFFAOYSA-N | |
| 6212 | |
| trichloromethane |
| CHCl3 | |
| MFCD00000826 | |
| trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
| CHEBI:35255 | |
| ClC(Cl)Cl |
Specifications
| -63°C | |
| Colorless | |
| 1.480g/cm³ | |
| 500 mL | |
| Glass Bottle | |
| 0.001% max. | |
| CHCl3 | |
| 213mbar at 20°C | |
| 0.02% max. | |
| HEDRZPFGACZZDS-UHFFFAOYSA-N | |
| trichloromethane | |
| 6212 | |
| 119.38 | |
| Certified ACS Reagent | |
| Chloroform, Approx. 0.75% ethanol as preservative |
| 67-66-3,64-17-5 | |
| 290°C | |
| 61°C | |
| Pass Test | |
| Liquid | |
| ≥99.8 % | |
| MFCD00000826 | |
| 0.56 mPa/s at 20°C | |
| trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
| ClC(Cl)Cl | |
| 119.37 | |
| CHEBI:35255 | |
| ≥99.8% | |
| 0.75% Ethanol |
Safety and Handling
WARNING!
Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. Irritating to eyes and skin. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. May be harmful if inhaled. May cause irritation of respiratory tract. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.
NFPA
Health:2
Flammability:1
Instability:0
DOTInformation : DOT Class 6.1, : Poison