Learn More
Chloroform (Ethanol as Preservative/Certified ACS), Fisher Chemical™
Meet or exceeds the latest ACS specifications. Actual lot of analysis on label
Supplier: Thermo Fisher Scientific BP29820
Specifications
| Chloroform | |
| Approx. 0.75% ethanol as preservative | |
| Colorless | |
| 1.480g/cm³ | |
| 20 L | |
| Steel Pail | |
| 0.001% max. | |
| CHCl3 | |
| 213mbar at 20°C | |
| 0.02% max. | |
| HEDRZPFGACZZDS-UHFFFAOYSA-N | |
| trichloromethane | |
| 6212 | |
| 119.38 | |
| Certified ACS Reagent |
| -63°C | |
| 67-66-3,64-17-5 | |
| 290°C | |
| 61°C | |
| Pass Test | |
| Liquid | |
| ≥99.8 % | |
| MFCD00000826 | |
| 0.56 mPa/s at 20°C | |
| trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
| ClC(Cl)Cl | |
| 119.37 | |
| CHEBI:35255 | |
| ≥99.8% | |
| 0.75% Ethanol |
Chemical Identifiers
| 67-66-3 | |
| 119.37 | |
| HEDRZPFGACZZDS-UHFFFAOYSA-N | |
| 6212 | |
| trichloromethane |
| CHCl3 | |
| MFCD00000826 | |
| trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
| CHEBI:35255 | |
| ClC(Cl)Cl |
Safety and Handling
WARNING!
Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. Irritating to eyes and skin. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. May be harmful if inhaled. May cause irritation of respiratory tract. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.
NFPA
Health:2
Flammability:1
Instability:0
DOTInformation : DOT Class 6.1, : Poison