missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chloroform (HPLC), Fisher Chemical™
HPLC mobile phase for analytical and preparative separations
Supplier: Fisher Chemical BPC6071
Description
HPLC mobile phase for analytical and preparative separations
Specifications
Chloroform | |
Approx. 50ppm Pentene as Preservative | |
0.01 AU max., 0.01 AU max. at 280nm, 0.05 AU max. at 270nm, 0.15 AU max. at 254nm, 0.15 AU max. at 260nm, 0.25 AU max. at 255nm, 0.32 AU max. at 250nm, 1.00 AU max. at 245nm, Pass Test | |
1.480g/cm³ | |
1 L | |
Glass Bottle | |
Mobile phase for HPLC applications | |
2ppm max. | |
CHCl3 | |
213mbar at 20°C | |
0.02% max. | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
trichloromethane | |
6212 | |
119.38 | |
HPLC |
-63°C | |
67-66-3,513-35-9 | |
Colorless | |
61°C | |
Pass Test | |
Liquid | |
1.4420 to 1.4460 (at 25°C) | |
99.8% min by GC | |
MFCD00000826 | |
0.56 mPa/s at 20°C | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
ClC(Cl)Cl | |
119.37 | |
CHEBI:35255 | |
≥99.8% | |
Approx. 50ppm Amylene as Preservative |
Chemical Identifiers
67-66-3 | |
119.37 | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
6212 | |
trichloromethane |
CHCl3 | |
MFCD00000826 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
CHEBI:35255 | |
ClC(Cl)Cl |
Safety and Handling
Emergency Overview
Harmful if swallowed. Irritating to eyes, respiratory system and skin. Symptoms of overexposure are dizziness, headache, tiredness, nausea, unconsciousness, cessation of breathing.
<special id
Signal Word: WARNING!
DOTInformation : DOT Class 6.1, : Poison