missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroform (HPLC), Fisher Chemical™

HPLC mobile phase for analytical and preparative separations

$768.50 - $1,324.75

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
C607-1
SDS View Documents
Thermo Fisher Scientific
BPC6071
1 L Glass Bottle N/A
 
C607-4
SDS Accessories View Documents
Thermo Fisher Scientific
C6074
4 L Glass Bottle N/A
 
C607SK-4
SDS View Documents
Thermo Fisher Scientific
BPC607SK4
4 L Safe-Cote™ Amber Glass N/A
 
Description

Description

HPLC mobile phase for analytical and preparative separations

Specifications

Chemical Identifiers

67-66-3
119.37
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
trichloromethane
CHCl3
MFCD00000826
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
CHEBI:35255
ClC(Cl)Cl

Specifications

-63°C
0.01 AU max., 0.01 AU max. at 280nm, 0.05 AU max. at 270nm, 0.15 AU max. at 254nm, 0.15 AU max. at 260nm, 0.25 AU max. at 255nm, 0.32 AU max. at 250nm, 1.00 AU max. at 245nm, Pass Test
1.480g/cm³
1 L
Glass Bottle
1.4420 to 1.4460 (at 25°C)
99.8% min by GC
MFCD00000826
0.56 mPa/s at 20°C
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
ClC(Cl)Cl
119.37
CHEBI:35255
≥99.8%
Approx. 50ppm Amylene as Preservative
67-66-3,513-35-9
Colorless
61°C
Pass Test
Liquid
2ppm max.
CHCl3
213mbar at 20°C
0.02% max.
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane
6212
119.38
Mobile phase for HPLC applications
Chloroform, Approx. 50ppm Pentene as Preservative
Videos
Safety and Handling

Safety and Handling

Emergency Overview
Harmful if swallowed. Irritating to eyes, respiratory system and skin. Symptoms of overexposure are dizziness, headache, tiredness, nausea, unconsciousness, cessation of breathing.
<special id

Signal Word: WARNING!

DOTInformation : DOT Class 6.1, : Poison

SDS
missing translation for 'documents'

missing translation for 'documents'