missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chlorosulfonyl isocyanate, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 162040500

 View more versions of this product

Catalog No. AC162040500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent, C-X bond formation reagent
Specifications

Specifications

Chlorosulfonyl isocyanate
-44.0°C
1.6260g/mL
>110°C
98+%
CClNO3S
ClSO2NCO
01,117; 02,70; 03,51; 04,90; 05,132; 06,122; 07,65; 08,105; 10,94; 11,125; 12,122; 13,80
chlorosulfonyl isocyanate, chlorosulfonylisocyanate, sulfuryl chloride isocyanate, n-carbonylsulfamyl chloride, chlorosulphonyl isocyanate, sulfurisocyanatidic chloride, n-chlorosulfonyl isocyanate, chlorosulfonyl imino methanone, csi, sulfurisocyanatidoyl chloride
WRJWRGBVPUUDLA-UHFFFAOYSA-N
N-(oxomethylidene)sulfamoyl chloride
70918
141.53
Crystalline Powder, Crystals or Granules
1189-71-5
White
106.0°C
Authentic
Glass bottle
1.4450 to 1.4490
50 mL
1.626
Solubility in water: reacts. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons,miscible with saturated and chlorinated,hydrocarbons
C(=NS(=O)(=O)Cl)=O
141.53
0.996 mPa.s (20°C)
98+%

Chemical Identifiers

1189-71-5
141.53
chlorosulfonyl isocyanate, chlorosulfonylisocyanate, sulfuryl chloride isocyanate, n-carbonylsulfamyl chloride, chlorosulphonyl isocyanate, sulfurisocyanatidic chloride, n-chlorosulfonyl isocyanate, chlorosulfonyl imino methanone, csi, sulfurisocyanatidoyl chloride
N-(oxomethylidene)sulfamoyl chloride
CClNO3S
WRJWRGBVPUUDLA-UHFFFAOYSA-N
70918
C(=NS(=O)(=O)Cl)=O
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 214-715-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only