missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chlorosulfonyl isocyanate, 98+%, Thermo Scientific Chemicals

$52.81 - $632.38

Chemical Identifiers

CAS 1189-71-5
Molecular Formula CClNO3S
Molecular Weight (g/mol) 141.53
InChI Key WRJWRGBVPUUDLA-UHFFFAOYSA-N
Synonym chlorosulfonyl isocyanate, chlorosulfonylisocyanate, sulfuryl chloride isocyanate, n-carbonylsulfamyl chloride, chlorosulphonyl isocyanate, sulfurisocyanatidic chloride, n-chlorosulfonyl isocyanate, chlorosulfonyl imino methanone, csi, sulfurisocyanatidoyl chloride
PubChem CID 70918
IUPAC Name N-(oxomethylidene)sulfamoyl chloride
SMILES C(=NS(=O)(=O)Cl)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC162040050
SDS View Documents
Thermo Scientific Chemicals
162040050
5 mL Glass bottle
Each for $52.81
 
AC162040500
SDS View Documents
Thermo Scientific Chemicals
162040500
50 mL Glass bottle
Each for $267.31
 
AC162042500
SDS View Documents
Thermo Scientific Chemicals
162042500
250 mL Glass bottle
Each for $632.38
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent, C-X bond formation reagent
Specifications

Chemical Identifiers

1189-71-5
141.53
chlorosulfonyl isocyanate, chlorosulfonylisocyanate, sulfuryl chloride isocyanate, n-carbonylsulfamyl chloride, chlorosulphonyl isocyanate, sulfurisocyanatidic chloride, n-chlorosulfonyl isocyanate, chlorosulfonyl imino methanone, csi, sulfurisocyanatidoyl chloride
N-(oxomethylidene)sulfamoyl chloride
CClNO3S
WRJWRGBVPUUDLA-UHFFFAOYSA-N
70918
C(=NS(=O)(=O)Cl)=O

Specifications

1189-71-5
White
106.0°C
Authentic
Glass bottle
1.4450 to 1.4490
5 mL
1.626
Solubility in water: reacts. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons,miscible with saturated and chlorinated,hydrocarbons
C(=NS(=O)(=O)Cl)=O
141.53
0.996 mPa.s (20°C)
98+%
Chlorosulfonyl isocyanate
-44.0°C
1.6260g/mL
>110°C
98+%
CClNO3S
ClSO2NCO
01,117; 02,70; 03,51; 04,90; 05,132; 06,122; 07,65; 08,105; 10,94; 11,125; 12,122; 13,80
chlorosulfonyl isocyanate, chlorosulfonylisocyanate, sulfuryl chloride isocyanate, n-carbonylsulfamyl chloride, chlorosulphonyl isocyanate, sulfurisocyanatidic chloride, n-chlorosulfonyl isocyanate, chlorosulfonyl imino methanone, csi, sulfurisocyanatidoyl chloride
WRJWRGBVPUUDLA-UHFFFAOYSA-N
N-(oxomethylidene)sulfamoyl chloride
70918
141.53
Crystalline Powder, Crystals or Granules
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 214-715-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only