missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chlorotriisopropylsilane, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 221260100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Chlorotriisopropylsilane | |
Colorless to Yellow | |
198.0°C | |
Authentic | |
Glass bottle | |
1.4520 to 1.4540 | |
MFCD00009656 | |
0.902 | |
Solubility in water: reacts | |
CC(C)[Si](Cl)(C(C)C)C(C)C | |
192.80 | |
192.81 | |
Liquid |
13154-24-0 | |
0.9020g/mL | |
63°C | |
96% min. (GC) | |
C9H21ClSi | |
[(CH3)2CH]3SiCl | |
10 g | |
triisopropylsilyl chloride, chlorotriisopropylsilane, triisopropylchlorosilane, chlorotris propan-2-yl silane, chloro triisopropyl silane, silane, chlorotris 1-methylethyl, tipscl, triisopropylsilylchloride, triisopropyl chlorosilane, triisopropylchlorosilane, redistilled | |
KQIADDMXRMTWHZ-UHFFFAOYSA-N | |
chloro-tri(propan-2-yl)silane | |
139400 | |
97% |
Chemical Identifiers
13154-24-0 | |
192.80 | |
KQIADDMXRMTWHZ-UHFFFAOYSA-N | |
139400 |
C9H21ClSi | |
MFCD00009656 | |
triisopropylsilyl chloride, chlorotriisopropylsilane, triisopropylchlorosilane, chlorotris propan-2-yl silane, chloro triisopropyl silane, silane, chlorotris 1-methylethyl, tipscl, triisopropylsilylchloride, triisopropyl chlorosilane, triisopropylchlorosilane, redistilled | |
CC(C)[Si](Cl)(C(C)C)C(C)C |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
RUO – Research Use Only