missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chlorotriisopropylsilane, 97%, Thermo Scientific Chemicals

$140.56 - $430.49

Chemical Identifiers

CAS 13154-24-0
Molecular Formula C9H21ClSi
Molecular Weight (g/mol) 192.80
MDL Number MFCD00009656
InChI Key KQIADDMXRMTWHZ-UHFFFAOYSA-N
Synonym triisopropylsilyl chloride, chlorotriisopropylsilane, triisopropylchlorosilane, chlorotris propan-2-yl silane, chloro triisopropyl silane, silane, chlorotris 1-methylethyl, tipscl, triisopropylsilylchloride, triisopropyl chlorosilane, triisopropylchlorosilane, redistilled
PubChem CID 139400
SMILES CC(C)[Si](Cl)(C(C)C)C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC221260100
SDS View Documents
Thermo Scientific Chemicals
221260100
10 g Glass bottle
Each for $140.56
 
AC221260500
SDS View Documents
Thermo Scientific Chemicals
221260500
50 g Glass bottle
Each for $430.49
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13154-24-0
192.80
KQIADDMXRMTWHZ-UHFFFAOYSA-N
139400
C9H21ClSi
MFCD00009656
triisopropylsilyl chloride, chlorotriisopropylsilane, triisopropylchlorosilane, chlorotris propan-2-yl silane, chloro triisopropyl silane, silane, chlorotris 1-methylethyl, tipscl, triisopropylsilylchloride, triisopropyl chlorosilane, triisopropylchlorosilane, redistilled
CC(C)[Si](Cl)(C(C)C)C(C)C

Specifications

13154-24-0
0.9020g/mL
63°C
96% min. (GC)
C9H21ClSi
[(CH3)2CH]3SiCl
10 g
triisopropylsilyl chloride, chlorotriisopropylsilane, triisopropylchlorosilane, chlorotris propan-2-yl silane, chloro triisopropyl silane, silane, chlorotris 1-methylethyl, tipscl, triisopropylsilylchloride, triisopropyl chlorosilane, triisopropylchlorosilane, redistilled
KQIADDMXRMTWHZ-UHFFFAOYSA-N
chloro-tri(propan-2-yl)silane
139400
97%
Chlorotriisopropylsilane
Colorless to Yellow
198.0°C
Authentic
Glass bottle
1.4520 to 1.4540
MFCD00009656
0.902
Solubility in water: reacts
CC(C)[Si](Cl)(C(C)C)C(C)C
192.80
192.81
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only